siRNA Details
| Gene Symbol: Irgc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014016.1 | NP_001014038.1 | 2 | 1013 | ||
| XM_006228425.2 | XP_006228487.1 | 2 | 1062 | |||
| XM_006228426.2 | XP_006228488.1 | 2 | 942 | |||
| XM_006228427.2 | XP_006228489.1 | 2 | 857 | |||
| XM_008758927.1 | XP_008757149.1 | 2 | 926 | |||
| GenBank mRNA | BC079034.1 | - | 2 | 960 | 7114307 | |
| BC079061.1 | - | 2 | 1013 | 7115757 | ||

Irgc
Genomic Map