siRNA Details
| Gene Symbol: Zfp574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024258.1 | NP_001019429.1 | 2 | 317 | ||
| XM_008758930.1 | XP_008757152.1 | 2 | 305 | |||
| XM_008758931.1 | XP_008757153.1 | 2 | 366 | |||
| XM_008758932.1 | XP_008757154.1 | 2 | 351 | |||
| GenBank mRNA | BC094953.1 | - | 2 | 317 | 7308498 | |

Zfp574
Genomic Map