siRNA Details
| Gene Symbol: Axl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013147.1 | NP_001013165.1 | 4 | 543 | ||
| NM_031794.1 | NP_113982.1 | 4 | 543 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Axl| Gene Symbol: Axl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013147.1 | NP_001013165.1 | 4 | 543 | ||
| NM_031794.1 | NP_113982.1 | 4 | 543 | |||