siRNA Details
| Gene Symbol: Adck4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012065.1 | NP_001012065.1 | 10 | 921 | ||
| GenBank mRNA | BC079227.1 | - | 10 | 921 | 7132453 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adck4| Gene Symbol: Adck4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012065.1 | NP_001012065.1 | 10 | 921 | ||
| GenBank mRNA | BC079227.1 | - | 10 | 921 | 7132453 | |