siRNA Details
Gene Symbol: Map3k10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191621.3 | NP_001178550.1 | 5 | 1636 | ||
XM_006228576.2 | XP_006228638.1 | 5 | 1845 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Map3k10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191621.3 | NP_001178550.1 | 5 | 1636 | ||
XM_006228576.2 | XP_006228638.1 | 5 | 1845 |