siRNA Details
| Gene Symbol: Dyrk1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107496.1 | NP_001100966.1 | 6 | 800 | ||
| XM_006228621.2 | XP_006228683.1 | 6 | 964 | |||
| XM_006228622.2 | XP_006228684.1 | 6 | 964 | |||
| XM_006228623.2 | XP_006228685.1 | 6 | 964 | |||
| XM_006228624.2 | XP_006228686.1 | 6 | 815 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001037957, NM_010092, NM_001271370, Y18280, AJ537610, and others.

Dyrk1b
Genomic Map