siRNA Details
| Gene Symbol: Zfp84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228671.2 | XP_006228733.1 | 3 | 6474 | ||
| XM_008759147.1 | XP_008757369.1 | 2 | 1738 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp84| Gene Symbol: Zfp84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228671.2 | XP_006228733.1 | 3 | 6474 | ||
| XM_008759147.1 | XP_008757369.1 | 2 | 1738 | |||