siRNA Details
| Gene Symbol: Ddx18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006996.1 | NP_001006997.1 | 10 | 1665 | ||
| GenBank mRNA | BC083919.1 | - | 10 | 1665 | 7135005 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddx18| Gene Symbol: Ddx18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006996.1 | NP_001006997.1 | 10 | 1665 | ||
| GenBank mRNA | BC083919.1 | - | 10 | 1665 | 7135005 | |