siRNA Details
| Gene Symbol: Wdr62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191623.2 | NP_001178552.2 | 8 | 1012 | ||
| XM_006228797.1 | XP_006228859.1 | 8 | 1074 | |||
| Gene Symbol: LOC103689973 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759279.1 | XP_008757501.1 | 7 | 1078 | ||

Wdr62
Genomic Map