siRNA Details
| Gene Symbol: Lrfn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107502.1 | NP_001100972.1 | 1 | 283 | ||
| XM_006228771.2 | XP_006228833.1 | 2 | 1298 | |||
| GenBank mRNA | BC158862.1 | - | 2 | 598 | 7933283 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrfn3| Gene Symbol: Lrfn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107502.1 | NP_001100972.1 | 1 | 283 | ||
| XM_006228771.2 | XP_006228833.1 | 2 | 1298 | |||
| GenBank mRNA | BC158862.1 | - | 2 | 598 | 7933283 | |