siRNA Details
Gene Symbol: RGD1563307 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_218493.7 | XP_218493.2 | 1 | 781 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1563307 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_218493.7 | XP_218493.2 | 1 | 781 |