siRNA Details
| Gene Symbol: Uba2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100579.1 | NP_001094049.1 | 8 | 867 | ||
| GenBank mRNA | BC161985.1 | - | 8 | 867 | 9030809 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uba2| Gene Symbol: Uba2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100579.1 | NP_001094049.1 | 8 | 867 | ||
| GenBank mRNA | BC161985.1 | - | 8 | 867 | 9030809 | |