siRNA Details
| Gene Symbol: Chst8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107504.2 | NP_001100974.1 | 2, 3 | 547 | ||
| XM_006228853.2 | XP_006228915.1 | 2, 3 | 1432 | |||
| XM_006228854.2 | XP_006228916.1 | 2, 3 | 858 | |||
| GenBank mRNA | BC161905.1 | - | 2, 3 | 547 | 8367390 | |

Chst8
Genomic Map