siRNA Details
| Gene Symbol: Rhpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107505.1 | NP_001100975.1 | 4, 5 | 426 | ||
| XM_006228887.2 | XP_006228949.1 | 4, 5 | 473 | |||
| XM_008759160.1 | XP_008757382.1 | 4, 5 | 473 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhpn2| Gene Symbol: Rhpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107505.1 | NP_001100975.1 | 4, 5 | 426 | ||
| XM_006228887.2 | XP_006228949.1 | 4, 5 | 473 | |||
| XM_008759160.1 | XP_008757382.1 | 4, 5 | 473 | |||