siRNA Details
Gene Symbol: Rhpn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107505.1 | NP_001100975.1 | 15 | 1877 | ||
XM_006228887.2 | XP_006228949.1 | 15 | 1924 | |||
XM_008759160.1 | XP_008757382.1 | 15 | 1924 |
