siRNA Details
| Gene Symbol: Nudt19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004258.1 | NP_001004258.1 | 2 | 840 | ||
| XM_008759161.1 | XP_008757383.1 | 3 | 1767 | |||
| GenBank mRNA | BC079088.1 | - | 2 | 840 | 7123358 | |
| FQ215228.1 | - | 2 | 935 | |||

Nudt19
Genomic Map