siRNA Details
| Gene Symbol: Vom2r37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099487.1 | NP_001092957.1 | 5 | 1639 | ||
| Gene Symbol: Vom2r38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099476.1 | NP_001092946.1 | 1858 | |||
More Information
Set Membership: |
|

Vom2r37
Genomic Map