siRNA Details
| Gene Symbol: Zfp819 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014017.2 | NP_001014039.1 | 5 | 574 | ||
| XM_006229044.2 | XP_006229106.1 | 6 | 591 | |||
| XM_006229045.2 | XP_006229107.1 | 7 | 2547 | |||
| XM_006229046.2 | XP_006229108.1 | 5 | 531 | |||
| XM_006229047.2 | XP_006229109.1 | 5 | 649 | |||
| XM_008759347.1 | XP_008757569.1 | 7 | 1729 | |||
| XM_008759348.1 | XP_008757570.1 | 8 | 7342 | |||
| XM_008759349.1 | XP_008757571.1 | 6 | 3015 | |||
| XM_008759350.1 | XP_008757572.1 | 4 | 325 | |||
| GenBank mRNA | BC087070.1 | - | 5 | 578 | 7134052 | |

Zfp819
Genomic Map