siRNA Details
| Gene Symbol: Lrrc4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271081.1 | NP_001258010.1 | 3 | 734 | ||
| XM_006229055.2 | XP_006229117.1 | 3 | 786 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc4b| Gene Symbol: Lrrc4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271081.1 | NP_001258010.1 | 3 | 734 | ||
| XM_006229055.2 | XP_006229117.1 | 3 | 786 | |||