siRNA Details
| Gene Symbol: Fuz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037646.1 | NP_001032735.1 | 2 | 323 | ||
| XM_006229059.2 | XP_006229121.1 | 2 | 362 | |||
| XM_006229060.2 | XP_006229122.1 | 2 | 362 | |||
| XM_006229061.2 | XP_006229123.1 | 2 | 363 | |||
| XM_008759373.1 | XP_008757595.1 | 2 | 363 | |||
| XM_008759374.1 | XP_008757596.1 | 2 | 363 | |||
| GenBank mRNA | BC107809.1 | - | 2 | 323 | 7453736 | |
| FQ228777.1 | - | 2 | 308 | |||

Fuz
Genomic Map