siRNA Details
| Gene Symbol: Ap2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107511.2 | NP_001100981.1 | 10, 11 | 1270 | ||
| XM_006229062.2 | XP_006229124.1 | 9, 10 | 3114 | |||
| XM_006229063.2 | XP_006229125.1 | 9, 10 | 3114 | |||
| XM_006229064.2 | XP_006229126.1 | 10, 11 | 1378 | |||
| XM_006229065.2 | XP_006229127.1 | 10, 11 | 1378 | |||

Ap2a1
Genomic Map