siRNA Details
Gene Symbol: Ccdc155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229199.2 | XP_006229261.1 | 12, 13 | 1601 | ||
XM_006229200.2 | XP_006229262.1 | 11, 12 | 1544 | |||
XM_006229201.2 | XP_006229263.1 | 12, 13 | 1290 |
