siRNA Details
| Gene Symbol: HIST1H4L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003546.2 | NP_003537.1 | 1 | 169 | ||
| GenBank mRNA | AB451459.1 | - | 1 | 168 | ||
| BC069392.1 | - | 1 | 169 | 7262681 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIST1H4L| Gene Symbol: HIST1H4L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003546.2 | NP_003537.1 | 1 | 169 | ||
| GenBank mRNA | AB451459.1 | - | 1 | 168 | ||
| BC069392.1 | - | 1 | 169 | 7262681 | ||