siRNA Details
| Gene Symbol: Sphk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012066.1 | NP_001012066.1 | 2 | 336 | ||
| XM_006229085.1 | XP_006229147.1 | 2 | 425 | |||
| XM_006229086.2 | XP_006229148.1 | 2 | 713 | |||
| XM_006229088.2 | XP_006229150.1 | 2 | 485 | |||
| XM_008759383.1 | XP_008757605.1 | 2 | 378 | |||
| XM_008759384.1 | XP_008757606.1 | 2 | 767 | |||
| XM_008759385.1 | XP_008757607.1 | 2 | 720 | |||
| GenBank mRNA | BC079120.1 | - | 2 | 336 | 7126682 | |
| FQ211663.1 | - | 2 | 502 | |||

Sphk2
Genomic Map