siRNA Details
| Gene Symbol: Tmem143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107513.1 | NP_001100983.1 | 5 | 671 | ||
| XM_006229089.2 | XP_006229151.1 | 5 | 666 | |||
| XM_006229090.2 | XP_006229152.1 | 4 | 532 | |||
| XM_006229092.2 | XP_006229154.1 | 5 | 637 | |||
| XM_008759386.1 | XP_008757608.1 | 5 | 637 | |||

Tmem143
Genomic Map