siRNA Details
| Gene Symbol: Ccdc114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126277.2 | NP_001119749.1 | 10 | 1204 | ||
| XM_008759387.1 | XP_008757609.1 | 11 | 3120 | |||
| XM_008759388.1 | XP_008757610.1 | 10 | 1456 | |||
| XM_008759389.1 | XP_008757611.1 | 7 | 778 | |||
| GenBank mRNA | BC160835.1 | - | 10 | 1206 | 7456475 | |

Ccdc114
Genomic Map