siRNA Details
| Gene Symbol: Ccdc114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126277.2 | NP_001119749.1 | 4 | 387 | ||
| XM_008759387.1 | XP_008757609.1 | 5 | 2303 | |||
| XM_008759388.1 | XP_008757610.1 | 4 | 639 | |||
| GenBank mRNA | BC160835.1 | - | 4 | 389 | 7456475 | |

Ccdc114
Genomic Map