siRNA Details
| Gene Symbol: Txlnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135859.1 | NP_001129331.1 | 10 | 1471 | ||
| XM_006227657.2 | XP_006227719.1 | 10 | 1456 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Txlnb| Gene Symbol: Txlnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135859.1 | NP_001129331.1 | 10 | 1471 | ||
| XM_006227657.2 | XP_006227719.1 | 10 | 1456 | |||