siRNA Details
| Gene Symbol: Hnrnph2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014019.1 | NP_001014041.1 | 4 | 1337 | ||
| XM_006257219.2 | XP_006257281.1 | 4 | 1380 | |||
| XM_006257220.2 | XP_006257282.1 | 3 | 1352 | |||
| XM_006257221.2 | XP_006257283.1 | 3 | 1336 | |||
| GenBank mRNA | BC079240.1 | - | 4 | 1337 | 7132657 | |

Hnrnph2
Genomic Map