siRNA Details
| Gene Symbol: Prame | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001055257.4 | XP_001055257.2 | 1, 2 | 100 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prame| Gene Symbol: Prame | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001055257.4 | XP_001055257.2 | 1, 2 | 100 | ||