siRNA Details
| Gene Symbol: Nipa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107518.1 | NP_001100988.1 | 7 | 1082 | ||
| XM_006229292.2 | XP_006229354.1 | 9 | 1185 | |||
| XM_006229295.2 | XP_006229357.1 | 7 | 896 | |||
| XM_006229296.2 | XP_006229358.1 | 7 | 952 | |||
| XM_006229297.2 | XP_006229359.1 | 6 | 941 | |||
| XM_006229298.2 | XP_006229360.1 | 6 | 759 | |||
| XM_006229299.2 | XP_006229361.1 | 6 | 831 | |||
| XM_008759390.1 | XP_008757612.1 | 6 | 787 | |||
| XM_008759391.1 | XP_008757613.1 | 6 | 763 | |||
| XM_008759392.1 | XP_008757614.1 | 8 | 1346 | |||
| GenBank mRNA | FQ222971.1 | - | 6 | 945 | ||

Nipa2
Genomic Map