siRNA Details
| Gene Symbol: Lrrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191624.1 | NP_001178553.1 | 16 | 2307 | ||
| XM_008759514.1 | XP_008757736.1 | 17 | 5021 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrk1| Gene Symbol: Lrrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191624.1 | NP_001178553.1 | 16 | 2307 | ||
| XM_008759514.1 | XP_008757736.1 | 17 | 5021 | |||