siRNA Details
| Gene Symbol: Agbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759581.1 | XP_008757803.1 | 1606 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Agbl1| Gene Symbol: Agbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759581.1 | XP_008757803.1 | 1606 | |||