siRNA Details
Gene Symbol: Mctp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191625.1 | NP_001178554.1 | 1 | 60 | ||
XM_006229375.2 | XP_006229437.1 | 2 | 409 | |||
XM_008759522.1 | XP_008757744.1 | 2 | 410 | |||
XM_008759523.1 | XP_008757745.1 | 2 | 738 | |||
XM_008759524.1 | XP_008757746.1 | 3 | 483 |
