siRNA Details
| Gene Symbol: Wdr73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107525.2 | NP_001100995.1 | 3 | 205 | ||
| XM_006229430.2 | XP_006229492.1 | 2 | 551 | |||
| GenBank mRNA | BC158658.1 | - | 3 | 205 | 8369176 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr73| Gene Symbol: Wdr73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107525.2 | NP_001100995.1 | 3 | 205 | ||
| XM_006229430.2 | XP_006229492.1 | 2 | 551 | |||
| GenBank mRNA | BC158658.1 | - | 3 | 205 | 8369176 | |