siRNA Details
| Gene Symbol: NME5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003551.2 | NP_003542.1 | 3 | 369 | ||
| XM_005272099.1 | XP_005272156.1 | 2 | 296 | |||
| XM_006714716.3 | XP_006714779.1 | 2 | 296 | |||
| XM_017009945.1 | XP_016865434.1 | 2 | 296 | |||
| GenBank mRNA | AF067724.1 | - | 3 | 339 | ||
| AK312181.1 | - | 3 | 316 | |||
| BC026182.2 | - | 3 | 341 | 5270050 | ||
| KU178416.1 | - | 2 | 291 | |||
| U90450.1 | - | 3 | 315 | |||
| Y14992.1 | - | 2 | 305 | |||

NME5
Genomic Map