siRNA Details
| Gene Symbol: Blm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748888.3 | XP_003748936.2 | 15, 16 | 3180 | ||
| XM_006229458.2 | XP_006229520.1 | 15, 16 | 3183 | |||
| XM_008759588.1 | XP_008757810.1 | 15, 16 | 3184 | |||
| XM_008759589.1 | XP_008757811.1 | 15, 16 | 3180 | |||
| XM_008759590.1 | XP_008757812.1 | 14, 15 | 2925 | |||
| XM_008759591.1 | XP_008757813.1 | 12, 13 | 2171 | |||

Blm
Genomic Map