siRNA Details
| Gene Symbol: Unc45a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037647.1 | NP_001032736.1 | 2 | 301 | ||
| XM_006229432.2 | XP_006229494.1 | 2 | 252 | |||
| GenBank mRNA | BC107919.1 | - | 2 | 301 | 7457637 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Unc45a| Gene Symbol: Unc45a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037647.1 | NP_001032736.1 | 2 | 301 | ||
| XM_006229432.2 | XP_006229494.1 | 2 | 252 | |||
| GenBank mRNA | BC107919.1 | - | 2 | 301 | 7457637 | |