siRNA Details
| Gene Symbol: Prc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107529.1 | NP_001100999.1 | 3 | 329 | ||
| XM_006229433.2 | XP_006229495.1 | 3 | 341 | |||
| XM_006229435.2 | XP_006229497.1 | 3 | 341 | |||
| XM_006229437.2 | XP_006229499.1 | 3 | 341 | |||

Prc1
Genomic Map