siRNA Details
| Gene Symbol: Gdpgp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229451.2 | XP_006229513.1 | 2 | 798 | ||
| XM_008759586.1 | XP_008757808.1 | 3 | 1151 | |||
| XM_218822.7 | XP_218822.2 | 3 | 927 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gdpgp1| Gene Symbol: Gdpgp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229451.2 | XP_006229513.1 | 2 | 798 | ||
| XM_008759586.1 | XP_008757808.1 | 3 | 1151 | |||
| XM_218822.7 | XP_218822.2 | 3 | 927 | |||