siRNA Details
| Gene Symbol: OR1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014565.2 | NP_055380.2 | 1 | 763 | ||
| GenBank mRNA | BC128248.1 | - | 1 | 763 | ||
| BC141935.1 | - | 1 | 763 | 40114774 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR1A1| Gene Symbol: OR1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014565.2 | NP_055380.2 | 1 | 763 | ||
| GenBank mRNA | BC128248.1 | - | 1 | 763 | ||
| BC141935.1 | - | 1 | 763 | 40114774 | ||