siRNA Details
| Gene Symbol: Det1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037194.1 | NP_001032271.1 | 2 | 382 | ||
| XM_006229398.2 | XP_006229460.1 | 2 | 394 | |||
| XM_008759536.1 | XP_008757758.1 | 2 | 394 | |||
| GenBank mRNA | BC100253.1 | - | 2 | 382 | 7456473 | |

Det1
Genomic Map