siRNA Details
| Gene Symbol: Fsd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191627.1 | NP_001178556.1 | 1 | 553 | ||
| XM_006229440.2 | XP_006229502.1 | 2 | 757 | |||
| XM_008759537.1 | XP_008757759.1 | 2 | 1974 | |||
| XM_008759538.1 | XP_008757760.1 | 3 | 1147 | |||
| GenBank mRNA | FQ223649.1 | - | 2 | 705 | ||

Fsd2
Genomic Map