siRNA Details
| Gene Symbol: RGD1310371 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100581.1 | NP_001094051.1 | 7 | 852 | ||
| GenBank mRNA | BC086444.1 | - | 6 | 714 | ||
| BC158775.1 | - | 7 | 852 | 7132188 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1310371| Gene Symbol: RGD1310371 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100581.1 | NP_001094051.1 | 7 | 852 | ||
| GenBank mRNA | BC086444.1 | - | 6 | 714 | ||
| BC158775.1 | - | 7 | 852 | 7132188 | ||