siRNA Details
| Gene Symbol: Mesdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008345.1 | NP_001008346.1 | 1 | 369 | ||
| GenBank mRNA | BC085892.1 | - | 1 | 369 | 7192668 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mesdc2| Gene Symbol: Mesdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008345.1 | NP_001008346.1 | 1 | 369 | ||
| GenBank mRNA | BC085892.1 | - | 1 | 369 | 7192668 | |