siRNA Details
| Gene Symbol: Usp35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759807.1 | XP_008758029.1 | 10, 11 | 2224 | ||
| XM_238899.7 | XP_238899.4 | 9, 10 | 2027 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp35| Gene Symbol: Usp35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759807.1 | XP_008758029.1 | 10, 11 | 2224 | ||
| XM_238899.7 | XP_238899.4 | 9, 10 | 2027 | |||