siRNA Details
| Gene Symbol: Tsku | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009965.2 | NP_001009965.1 | 2 | 579 | ||
| GenBank mRNA | AY533242.1 | - | 2 | 500 | ||
| BC100644.1 | - | 2 | 579 | 7930712 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tsku| Gene Symbol: Tsku | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009965.2 | NP_001009965.1 | 2 | 579 | ||
| GenBank mRNA | AY533242.1 | - | 2 | 500 | ||
| BC100644.1 | - | 2 | 579 | 7930712 | ||