siRNA Details
| Gene Symbol: Fchsd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107539.1 | NP_001101009.1 | 7 | 636 | ||
| XM_008759720.1 | XP_008757942.1 | 8 | 828 | |||
| XM_008759721.1 | XP_008757943.1 | 8 | 828 | |||
| XM_008759722.1 | XP_008757944.1 | 8 | 828 | |||

Fchsd2
Genomic Map