siRNA Details
| Gene Symbol: Lamtor1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199102.1 | NP_954533.1 | 5 | 501 | ||
| GenBank mRNA | BC061783.1 | - | 5 | 501 | 5623933 | |
| FQ214531.1 | - | 5 | 505 | |||
| FQ219988.1 | - | 5 | 514 | |||
| FQ220581.1 | - | 5 | 497 | |||
| FQ220693.1 | - | 5 | 499 | |||

Lamtor1
Genomic Map