siRNA Details
| Gene Symbol: Rnf121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107540.2 | NP_001101010.1 | 7 | 730 | ||
| XM_006229878.2 | XP_006229940.1 | 7 | 955 | |||
| XM_006229879.2 | XP_006229941.1 | 8 | 1073 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf121| Gene Symbol: Rnf121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107540.2 | NP_001101010.1 | 7 | 730 | ||
| XM_006229878.2 | XP_006229940.1 | 7 | 955 | |||
| XM_006229879.2 | XP_006229941.1 | 8 | 1073 | |||